accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFFFFFFFFF

CAS RN 81190-28-5

IUPAC Name: 2-(2,2,3,3,4,4,5,5,5-nonafluoropentyl)oxirane
Synonyms: 2-(2,2,3,3,4,4,5,5,5-nonafluoropentyl)oxirane
Molecular Weight (g/mol): 276.1
Molecular Formula: C7H5F9O
InChi Key: WUKHWLIEBSRTRH-UHFFFAOYNA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)CC1CO1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results