accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OOHN

CAS RN 81282-60-2

IUPAC Name: 7-amino-2,3-dihydro-1,3-benzoxazol-2-one
Synonyms: 7-amino-3H-1,3-benzoxazol-2-one
Molecular Weight (g/mol): 150.14
Molecular Formula: C7H6N2O2
InChi Key: CLCPWTXGFUIRJE-UHFFFAOYSA-N
SMILES: NC1=C2OC(=O)NC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)