accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFOFFFFFFFFFF

CAS RN 813-44-5

IUPAC Name: 1,1,1,2,4,5,5,5-octafluoro-2,4-bis(trifluoromethyl)pentan-3-one
Synonyms: 1,1,1,2,4,5,5,5-octafluoro-2,4-bis(trifluoromethyl)pentan-3-one
Molecular Weight (g/mol): 366.05
Molecular Formula: C7F14O
InChi Key: GRVMOMUDALILLH-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(C(=O)C(F)(C(F)(F)F)C(F)(F)F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results