accessibility menu, dialog, popup

UserName

Viewing profile as user:
BiOHOOOOOO

CAS RN 813-93-4

IUPAC Name: bismuth(3+) 2-hydroxypropane-1,2,3-tricarboxylate
Synonyms: bismuth citrate
Molecular Weight (g/mol): 398.08
Molecular Formula: C6H5BiO7
InChi Key: ANERHPOLUMFRDC-UHFFFAOYSA-K
SMILES: [Bi+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (5)
  • (5)
  • (2)
Physical Form 
  • (2)
  • (5)
  • (5)
Grade 
  • (5)
  • (5)
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (10)
  • (2)
  • (10)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (5)
  • (5)
  • (5)
  • (2)
  • (5)
  • (2)