accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CH3CCH3ONHOHO

CAS RN 81335-77-5

IUPAC Name: 5-ethyl-2-[4-methyl-5-oxo-4-(propan-2-yl)-4,5-dihydro-1H-imidazol-2-yl]pyridine-3-carboxylic acid
Synonyms: imazethapyr
Molecular Weight (g/mol): 289.34
Molecular Formula: C15H19N3O3
InChi Key: XVOKUMIPKHGGTN-UHFFFAOYNA-N
SMILES: CCC1=CN=C(C2=NC(C)(C(C)C)C(=O)N2)C(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results