accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 81380-24-7

H3CNNNCH3H3C(E/Z)

CAS RN 81380-24-7

IUPAC Name: 9-ethyl-3-{[2-ethyl-2-(3-methylphenyl)hydrazin-1-ylidene]methyl}-9H-carbazole
Synonyms: 9-ethyl-3-{[2-ethyl-2-(3-methylphenyl)hydrazin-1-ylidene]methyl}carbazole
Molecular Weight (g/mol): 355.49
Molecular Formula: C24H25N3
InChi Key: VIRJEGJEGJEEDN-UHFFFAOYSA-N
SMILES: CCN(N=CC1=CC2=C(C=C1)N(CC)C1=C2C=CC=C1)C1=CC=CC(C)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results