accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HSHS

CAS RN 814-67-5

IUPAC Name: propane-1,2-dithiol
Synonyms: 1,2-propanedithiol
Molecular Weight (g/mol): 108.22
Molecular Formula: C3H8S2
InChi Key: YGKHJWTVMIMEPQ-UHFFFAOYNA-N
SMILES: CC(S)CS
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results