accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 814-78-8

IUPAC Name: 3-methylbut-3-en-2-one
Synonyms: 3-buten-2-one, 3-methyl-
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: ZGHFDIIVVIFNPS-UHFFFAOYSA-N
SMILES: CC(=C)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results