accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CONNH2NOCH3NCH3NHOO

CAS RN 81403-68-1

IUPAC Name: hydrogen N-{3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)(methyl)amino]propyl}oxolane-2-carboxamide chloride
Synonyms: uroxatral alfuzosin hydrochloride
Molecular Weight (g/mol): 425.91
Molecular Formula: C19H28ClN5O4
InChi Key: YTNKWDJILNVLGX-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N(C)CCCNC(=O)C1CCCO1
Grade 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)