accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OON

CAS RN 81477-94-3

IUPAC Name: tert-butyl 2-[(diphenylmethylidene)amino]acetate
Synonyms: tert-butyl 2-[(diphenylmethylidene)amino]acetate
Molecular Weight (g/mol): 295.38
Molecular Formula: C19H21NO2
InChi Key: YSHDPXQDVKNPKA-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CN=C(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)