accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COCH3CH3H3C

CAS RN 815-24-7

IUPAC Name: 2,2,4,4-tetramethylpentan-3-one
Synonyms: 2,2,4,4-tetramethylpentan-3-one
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: UIQGEWJEWJMQSL-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)C(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results