CAS RN 815-52-1
IUPAC Name:
2-bromopentan-3-one
Synonyms:
2-bromopentan-3-one
Molecular Weight (g/mol):
165.03
Molecular Formula:
C5H9BrO
InChi Key:
VUDTYIUNUSPULX-UHFFFAOYNA-N
SMILES:
CCC(=O)C(C)Br
1
–
1
of
1
results
1
2-Bromo-3-pentanone 95.0+%, TCI America™
CAS: 815-52-1 Molecular Formula: C5H9BrO Molecular Weight (g/mol): 165.03 MDL Number: MFCD11036312 InChI Key: VUDTYIUNUSPULX-UHFFFAOYSA-N PubChem CID: 545092 IUPAC Name: 2-bromopentan-3-one SMILES: CCC(=O)C(C)Br