accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 81549-07-7

H3CCH3NOSOOCH3CH3H3C

CAS RN 81549-07-7

IUPAC Name: (propan-2-ylidene)amino 2,4,6-trimethylbenzene-1-sulfonate
Synonyms: propan-2-ylideneamino 2,4,6-trimethylbenzenesulfonate
Molecular Weight (g/mol): 255.33
Molecular Formula: C12H17NO3S
InChi Key: BCPWUOSBRCQZFB-UHFFFAOYSA-N
SMILES: CC(C)=NOS(=O)(=O)C1=C(C)C=C(C)C=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)