accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 815631-56-2

IUPAC Name: 2-(4-fluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(4-fluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 236.09
Molecular Formula: C13H18BFO2
InChi Key: HHWOQSZBVGPYMH-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC=C1B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results