accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOBr

CAS RN 816-39-7

IUPAC Name: 1,3-dibromopropan-2-one
Synonyms: 2-propanone, 1,3-dibromo-
Molecular Weight (g/mol): 215.87
Molecular Formula: C3H4Br2O
InChi Key: LQQKDSXCDXHLLF-UHFFFAOYSA-N
SMILES: BrCC(=O)CBr
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)