accessibility menu, dialog, popup

UserName

Viewing profile as user:
SBrBrSOOCl

CAS RN 81606-31-7

IUPAC Name: 4,5-dibromothiophene-2-sulfonyl chloride
Synonyms: 4,5-dibromothiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 340.43
Molecular Formula: C4HBr2ClO2S2
InChi Key: WJYGHWXWQSCONR-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC(Br)=C(Br)S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results