accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOHHNONHOHNONHOOHNH2CH3OHO

CAS RN 81678-16-2

IUPAC Name: 2-[2-(2-{2-[2-amino-3-(4-hydroxyphenyl)propanamido]acetamido}acetamido)-3-phenylpropanamido]-4-methylpentanoic acid; acetic acid
Synonyms: acetic acid; tyr-gly-gly-phe-leu
Molecular Weight (g/mol): 615.68
Molecular Formula: C30H41N5O9
InChi Key: BAPJLGTVLSUVBN-UHFFFAOYNA-N
SMILES: CC(O)=O.CC(C)CC(NC(=O)C(CC1=CC=CC=C1)NC(=O)CNC(=O)CNC(=O)C(N)CC1=CC=C(O)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)