accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NFFF

CAS RN 819-46-5

IUPAC Name: 4,4,4-trifluorobutan-1-amine
Synonyms: 4,4,4-trifluorobutan-1-amine
Molecular Weight (g/mol): 127.11
Molecular Formula: C4H8F3N
InChi Key: LAXWLCVPJLBABV-UHFFFAOYSA-N
SMILES: NCCCC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)