accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 819050-88-9

IUPAC Name: 3-[3-(2-carboxy-2,2-dimethylethyl)phenyl]-2,2-dimethylpropanoic acid
Synonyms: 3-[3-(2-carboxy-2,2-dimethylethyl)phenyl]-2,2-dimethylpropanoic acid
Molecular Weight (g/mol): 278.35
Molecular Formula: C16H22O4
InChi Key: BZWQVGTVYGEXTE-UHFFFAOYSA-N
SMILES: CC(C)(CC1=CC(CC(C)(C)C(O)=O)=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results