CAS RN 819849-20-2
IUPAC Name:
(2-fluoro-5-nitrophenyl)boronic acid
Synonyms:
2-fluoro-5-nitrophenylboronic acid
Molecular Weight (g/mol):
184.92
Molecular Formula:
C6H5BFNO4
InChi Key:
SYAFEPQKPQIRAL-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(=CC=C1F)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
2-Fluoro-5-nitrobenzeneboronic acid, 97%
2-Fluoro-5-nitrobenzeneboronic acid, 97%
CAS: 819849-20-2 Molecular Formula: C6H5BFNO4 Molecular Weight (g/mol): 184.92 MDL Number: MFCD03092929 InChI Key: SYAFEPQKPQIRAL-UHFFFAOYSA-N Synonym: 2-fluoro-5-nitrophenyl boronic acid,2-fluoro-5-nitrophenylboronicacid,2-fluoro-5-nitrobenzeneboronic acid,5-nitro-2-fluorophenylboronic acid,boronic acid, 2-fluoro-5-nitrophenyl,2-fluoro-5-nitro-phenyl boronic acid,acmc-209pmq,ksc649e7n,2-fluoro-5-nitro phenyl boronic acid,b-2-fluoro-5-nitrophenyl-boronic acid PubChem CID: 2763280 IUPAC Name: (2-fluoro-5-nitrophenyl)boronic acid SMILES: OB(O)C1=CC(=CC=C1F)[N+]([O-])=O