accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 81998-03-0

IUPAC Name: 4,4'-dimethyl-[2,2'-bipyridin]-1-ium-1-olate
Synonyms: 4,4'-dimethyl-[2,2'-bipyridin]-1-ium-1-olate
Molecular Weight (g/mol): 200.24
Molecular Formula: C12H12N2O
InChi Key: VNSKKRAUVGCMPH-UHFFFAOYSA-N
SMILES: CC1=CC(=NC=C1)C1=[N+]([O-])C=CC(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results