CAS RN 82-22-4
CAS RN 82-22-4
IUPAC Name:
1-[(9,10-dioxo-9,10-dihydroanthracen-1-yl)amino]-9,10-dihydroanthracene-9,10-dione
Synonyms:
anthrimide
Molecular Weight (g/mol):
429.43
Molecular Formula:
C28H15NO4
InChi Key:
WITKIIIPSSFHST-UHFFFAOYSA-N
SMILES:
O=C1C2=CC=CC=C2C(=O)C2=C(NC3=CC=CC4=C3C(=O)C3=CC=CC=C3C4=O)C=CC=C12