accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82-34-8

IUPAC Name: 1-nitro-9,10-dihydroanthracene-9,10-dione
Synonyms: 1-nitroanthraquinone
Molecular Weight (g/mol): 253.21
Molecular Formula: C14H7NO4
InChi Key: YCANAXVBJKNANM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC2=C1C(=O)C1=CC=CC=C1C2=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)