accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOO

CAS RN 82-44-0

IUPAC Name: 1-chloro-9,10-dihydroanthracene-9,10-dione
Synonyms: 1-chloroanthraquinone
Molecular Weight (g/mol): 242.66
Molecular Formula: C14H7ClO2
InChi Key: BOCJQSFSGAZAPQ-UHFFFAOYSA-N
SMILES: ClC1=CC=CC2=C1C(=O)C1=CC=CC=C1C2=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)