accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OO

CAS RN 82-45-1

IUPAC Name: 1-amino-9,10-dihydroanthracene-9,10-dione
Synonyms: 1-aminoanthraquinone
Molecular Weight (g/mol): 223.23
Molecular Formula: C14H9NO2
InChi Key: KHUFHLFHOQVFGB-UHFFFAOYSA-N
SMILES: NC1=CC=CC2=C1C(=O)C1=CC=CC=C1C2=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)