accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 82-86-0

IUPAC Name: 1,2-dihydroacenaphthylene-1,2-dione
Synonyms: acenaphthoquinone 1,2-acenaphthenequinone acenaphthylene-1,2-quinone diketoacenaphthene acenaphthaquinone acenaphthylenedione acenaphthenedione acenaphthodione
Molecular Weight (g/mol): 182.18
Molecular Formula: C12H6O2
InChi Key: AFPRJLBZLPBTPZ-UHFFFAOYSA-N
SMILES: O=C1C(=O)C2=C3C1=CC=CC3=CC=C2
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)