accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 820-10-0

IUPAC Name: 2-amino-4-methanesulfonylbutanoic acid
Synonyms: methioninesulfone
Molecular Weight (g/mol): 181.21
Molecular Formula: C5H11NO4S
InChi Key: UCUNFLYVYCGDHP-UHFFFAOYNA-N
SMILES: CS(=O)(=O)CCC(N)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)