accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrO

CAS RN 82072-23-9

IUPAC Name: 3-(bromomethyl)benzaldehyde
Synonyms: 3-(bromomethyl)benzaldehyde
Molecular Weight (g/mol): 199.05
Molecular Formula: C8H7BrO
InChi Key: OEPGAYXSRGROSQ-UHFFFAOYSA-N
SMILES: BrCC1=CC=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results