accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 821-10-3

IUPAC Name: 1,4-dichlorobut-2-yne
Synonyms: 2-butyne, 1,4-dichloro-
Molecular Weight (g/mol): 122.98
Molecular Formula: C4H4Cl2
InChi Key: RCHDLEVSZBOHOS-UHFFFAOYSA-N
SMILES: ClCC#CCCl
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)