accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOH(E)

CAS RN 821-11-4

IUPAC Name: (2E)-but-2-ene-1,4-diol
Synonyms: 2-butene-1,4-diol (trans)
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: ORTVZLZNOYNASJ-OWOJBTEDSA-N
SMILES: OC\C=C\CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results