accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHO

CAS RN 821-38-5

IUPAC Name: tetradecanedioic acid
Synonyms: tetradecanedioic acid
Molecular Weight (g/mol): 258.36
Molecular Formula: C14H26O4
InChi Key: HQHCYKULIHKCEB-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCCCCC(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)