accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 821-99-8

IUPAC Name: 1,9-dichlorononane
Synonyms: nonane, 1,9-dichloro-
Molecular Weight (g/mol): 197.14
Molecular Formula: C9H18Cl2
InChi Key: JMGRNJZUQCEJDB-UHFFFAOYSA-N
SMILES: ClCCCCCCCCCCl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)