accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3H3CNSOOFFFSOOFFF

CAS RN 82113-66-4

IUPAC Name: 1,1,1-trifluoro-N-trifluoromethanesulfonyl-N-(trimethylsilyl)methanesulfonamide
Synonyms: 1,1,1-trifluoro-N-trifluoromethanesulfonyl-N-(trimethylsilyl)methanesulfonamide
Molecular Weight (g/mol): 353.33
Molecular Formula: C5H9F6NO4S2Si
InChi Key: MLIRNWUYOYIGBZ-UHFFFAOYSA-N
SMILES: C[Si](C)(C)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results