accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82156-58-9

IUPAC Name: ethyl 2-(1-benzofuran-3-yl)acetate
Synonyms: ethyl 2-(1-benzofuran-3-yl)acetate
Molecular Weight (g/mol): 204.23
Molecular Formula: C12H12O3
InChi Key: LSGDCGDXQLDKGO-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=COC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results