CAS RN 822-08-2
IUPAC Name:
undecane-1,11-diamine
Synonyms:
undecane-1,11-diamine
Molecular Weight (g/mol):
186.34
Molecular Formula:
C11H26N2
InChi Key:
KLNPWTHGTVSSEU-UHFFFAOYSA-N
SMILES:
NCCCCCCCCCCCN
1
–
1
of
1
results
1
1,11-Diaminoundecane 98.0+%, TCI America™
CAS: 822-08-2 Molecular Formula: C11H26N2 Molecular Weight (g/mol): 186.34 MDL Number: MFCD00142358 InChI Key: KLNPWTHGTVSSEU-UHFFFAOYSA-N Synonym: Undecamethylenediamine, 1,11-Undecanediamine PubChem CID: 69966 IUPAC Name: undecane-1,11-diamine SMILES: NCCCCCCCCCCCN