accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3COO

CAS RN 822-12-8

IUPAC Name: sodium tetradecanoate
Synonyms: sodium myristate
Molecular Weight (g/mol): 250.36
Molecular Formula: C14H27NaO2
InChi Key: JUQGWKYSEXPRGL-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCCCCCC([O-])=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)