accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCl

CAS RN 822-13-9

IUPAC Name: 1-chlorotridecane
Synonyms: 1-chlorotridecane
Molecular Weight (g/mol): 218.81
Molecular Formula: C13H27Cl
InChi Key: ASZMYJSJEOGSBR-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results