accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3OO(E)(E)

CAS RN 822-17-3

IUPAC Name: sodium (9E,12E)-octadeca-9,12-dienoate
Synonyms: sodium linoleate
Molecular Weight (g/mol): 302.43
Molecular Formula: C18H31NaO2
InChi Key: WYPBVHPKMJYUEO-JRYFSKGNSA-M
SMILES: [Na+].CCCCC\C=C\C\C=C\CCCCCCCC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)