CAS RN 82223-91-4
CAS RN 82223-91-4
IUPAC Name:
trimethyl[(2-methylpenta-1,3-dien-1-yl)oxy]silane
Synonyms:
trimethyl[(2-methylpenta-1,3-dien-1-yl)oxy]silane
Molecular Weight (g/mol):
170.33
Molecular Formula:
C9H18OSi
InChi Key:
KCWJPELNBSKTNR-UHFFFAOYSA-N
SMILES:
CC=CC(C)=CO[Si](C)(C)C
1
–
1
of
1
results
1
Trimethyl[[(1E,3E)-2-methylpenta-1,3-dien-1-yl]oxy]silane 95.0+%, TCI America™
CAS: 82223-91-4 Molecular Formula: C9H18OSi Molecular Weight (g/mol): 170.327 InChI Key: KCWJPELNBSKTNR-UHFFFAOYSA-N Synonym: (1E,3E)-2-Methyl-1-(trimethylsilyloxy)-1,3-pentadiene PubChem CID: 71407348 IUPAC Name: trimethyl(2-methylpenta-1,3-dienoxy)silane SMILES: CC=CC(=CO[Si](C)(C)C)C