CAS RN 82297-90-3
CAS RN 82297-90-3
IUPAC Name:
bromo(3,5-dichlorophenyl)magnesium
Synonyms:
bromo(3,5-dichlorophenyl)magnesium
Molecular Weight (g/mol):
250.2
Molecular Formula:
C6H3BrCl2Mg
InChi Key:
PWOVECVCCCTJFQ-UHFFFAOYSA-M
SMILES:
ClC1=CC([Mg]Br)=CC(Cl)=C1
1
–
1
of
1
results
1
Sigma Aldrich 3-Chloro-4-Fluorophenylmagnesium Bromide Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.