accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 823-36-9

IUPAC Name: 2-ethylcyclopentane-1,3-dione
Synonyms: 2-ethylcyclopentane-1,3-dione
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: YDFBIBUYOUFJMR-UHFFFAOYSA-N
SMILES: CCC1C(=O)CCC1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)