accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNN

CAS RN 823188-79-0

IUPAC Name: methyl({[4-(pyrrolidin-1-yl)phenyl]methyl})amine
Synonyms: methyl({[4-(pyrrolidin-1-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 190.29
Molecular Formula: C12H18N2
InChi Key: DJSIBXGPJNBMBD-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(C=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results