accessibility menu, dialog, popup

UserName

Viewing profile as user:
SSNH

CAS RN 82358-09-6

IUPAC Name: 2,3-dihydro-1,3-thiazole-2-thione
Synonyms: 3H-1,3-thiazole-2-thione
Molecular Weight (g/mol): 117.18
Molecular Formula: C3H3NS2
InChi Key: OCVLSHAVSIYKLI-UHFFFAOYSA-N
SMILES: S=C1NC=CS1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)