accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 824-11-3

IUPAC Name: 4-ethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane
Synonyms: trimethylolpropane phosphite
Molecular Weight (g/mol): 162.13
Molecular Formula: C6H11O3P
InChi Key: QRUSNTDXJQBKBI-UHFFFAOYSA-N
SMILES: CCC12COP(OC1)OC2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results