accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHHO

CAS RN 824-46-4

IUPAC Name: 2-methoxybenzene-1,4-diol
Synonyms: 1,4-benzenediol, 2-methoxy-
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: LAQYHRQFABOIFD-UHFFFAOYSA-N
SMILES: COC1=CC(O)=CC=C1O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)