accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 824-94-2

IUPAC Name: 1-(chloromethyl)-4-methoxybenzene
Synonyms: 4-methoxybenzyl chloride
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: MOHYOXXOKFQHDC-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCl)C=C1
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)