accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSNNONHHCl

CAS RN 824958-12-5

IUPAC Name: 6-{4-[2-(methylsulfanyl)phenyl]piperazin-1-yl}-N-(1,2,3,4-tetrahydronaphthalen-1-yl)hexanamide hydrochloride
Synonyms: 6-{4-[2-(methylsulfanyl)phenyl]piperazin-1-yl}-N-(1,2,3,4-tetrahydronaphthalen-1-yl)hexanamide hydrochloride
Molecular Weight (g/mol): 488.13
Molecular Formula: C27H38ClN3OS
InChi Key: DWGKCWWWKHCVDH-UHFFFAOYNA-N
SMILES: Cl.CSC1=CC=CC=C1N1CCN(CCCCCC(=O)NC2CCCC3=CC=CC=C23)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results