accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHO(S)INHOO

CAS RN 82565-68-2

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-iodophenyl)propanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-iodophenyl)propanoic acid|FMOC-4-IODO-L-PHENYLALANINE
Molecular Weight (g/mol): 513.33
Molecular Formula: C24H20INO4
InChi Key: LXOXXTQKKRJNNB-QFIPXVFZSA-N
SMILES: OC(=O)[C@H](CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)