accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 826-35-7

IUPAC Name: 1,2-dimethyl (1R,2R)-cyclopropane-1,2-dicarboxylate
Synonyms: 1,2-dimethyl (1R,2R)-cyclopropane-1,2-dicarboxylate
Molecular Weight (g/mol): 158.15
Molecular Formula: C7H10O4
InChi Key: JBVOSZYUSFDYIN-RFZPGFLSSA-N
SMILES: COC(=O)[C@@H]1C[C@H]1C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results