accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 826-55-1

IUPAC Name: 2-methyl-2-phenylpropanoic acid
Synonyms: cuminic acid
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: YYEROYLAYAVZNW-UHFFFAOYSA-N
SMILES: CC(C)(C(O)=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
Boiling Point 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)